missing translation for 'onlineSavingsMsg'
Learn More

(Methoxycarbonylsulfamoyl)triethylammonium Hydroxide Inner Salt 97.0+%, TCI America™

Catalog No. p-7132639
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12791G 1 g
M12795G 5 g
M127925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M12791G Supplier TCI America Supplier No. M12791G
Only null left

Chemical Identifiers

CAS 29684-56-8
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00077815
InChI Key YSHOWEKUVWPFNR-UHFFFAOYSA-N
Synonym methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide
PubChem CID 11032497
IUPAC Name (methoxycarbonyl)[(triethylazaniumyl)sulfonyl]azanide
SMILES CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OC

Specifications

Melting Point 79°C
Quantity 1 g
Formula Weight 238.30
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (Methoxycarbonylsulfamoyl)triethylammonium Hydroxide Inner Salt
TSCA No
Recommended Storage Refrigerator