missing translation for 'onlineSavingsMsg'
Learn More

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific™

Numéro de catalogue. AC294510250
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
25g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC294510250 25g Glass bottle
AC294510010 1g Glass bottle
AC294510050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC294510250 Fournisseur Thermo Scientific Chemicals Code fournisseur 294510250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 29684-56-8
Formule moléculaire C8H18N2O4S
Poids moléculaire (g/mol) 238.30
Numéro MDL MFCD00077815,MFCD00077815,MFCD00077815
Clé InChI YSHOWEKUVWPFNR-UHFFFAOYSA-N
Synonyme methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide
PubChem CID 11032497
Nom de l’IUPAC triethyl({[(methoxycarbonyl)imino](oxido)oxo-λ⁶-sulfanyl})azanium
SOURIRES CC[N+](CC)(CC)S([O-])(=O)=NC(=O)OC

Spécifications

Nom ou substance chimique (Methoxycarbonylsulfamoyl)triethylammonium hydroxide
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96% min
Emballage Glass bottle
Formule linéaire CH3O2CNSO2N(C2H5)3
Quantité 25g
Fieser 14,208
Indice de Merck 15,1505
Poids de formule 238.31
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only