missing translation for 'onlineSavingsMsg'
Learn More

Methoxybenzoquinone 99.0+%, TCI America™

Catalog No. p-7132511
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M10451G 1 g
M10455G 5 g
2 options

Catalog No. M10451G

Supplier: TCI America M10451G

Only null left

Chemical Identifiers

CAS 2880-58-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00144755
InChI Key ZJKWJHONFFKJHG-UHFFFAOYSA-N
PubChem CID 76146
IUPAC Name 2-methoxycyclohexa-2,5-diene-1,4-dione
SMILES COC1=CC(=O)C=CC1=O

Specifications

Melting Point 147°C
Color Yellow
Quantity 1 g
Formula Weight 138.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methoxybenzoquinone