missing translation for 'onlineSavingsMsg'
Learn More

Methoxyamine Hydrochloride, MP Biomedicals™

Catalog No. p-3547203
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15540525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15540525 Supplier MP Biomedicals Inc Supplier No. 0215540525
Only null left

Methoxyamine Hydrochloride is used as a reagent for the preparation of O-methyl oximes.

  • Also used to make O-methyl oximes from aldehydes, or ketones.

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point 300°F to 304°F (decomposes)
Quantity 25 g
Solubility Information Soluble
Formula Weight 83.52
Physical Form Crystalline Powder
Chemical Name or Material Methoxyamine Hydrochloride
RTECSNumber NC3980000
Recommended Storage Room Temperature

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.