missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Methoxyamine hydrochloride
Description
Methoxyamine hydrochloride
Specifications
Specifications
| CAS | 593-56-6 |
| Melting Point | 151°C to 154°C (lit.) |
| Molecular Formula | CH5NO -+ HCl |
| Linear Formula | CH3ONH2 -+ HCl |
| MDL Number | MFCD00012951 |
| Quantity | 25 g |
| Synonym | O-Methylhydroxylamine hydrochloride; Methoxylamine hydrochloride |
| Molecular Weight (g/mol) | 83.52 |
| Percent Purity | 98% |