missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Methoxyamine hydrochloride

Catalog No. 226904100G
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
226904100G 100 g
22690425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 226904100G Supplier Sigma Aldrich Supplier No. 226904100G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methoxyamine hydrochloride

Specifications

CAS 593-56-6
Melting Point 151°C to 154°C (lit.)
Molecular Formula CH5NO -+ HCl
Linear Formula CH3ONH2 -+ HCl
MDL Number MFCD00012951
Quantity 100 g
Synonym O-Methylhydroxylamine hydrochloride; Methoxylamine hydrochloride
Molecular Weight (g/mol) 83.52
Percent Purity 98%