missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetyl chloride, 97%, stabilized

CAS: 38870-89-2 | C3H5ClO2 | 108.52 g/mol

$589.91 - $589.91

Chemical Identifiers

CAS 38870-89-2
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00000728
InChI Key JJKWHOSQTYYFAE-UHFFFAOYSA-N
Synonym methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
PubChem CID 96623
IUPAC Name 2-methoxyacetyl chloride
SMILES COCC(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125700500
SDS View Documents
Thermo Scientific Chemicals
125700500
50 g Glass bottle
Each for $589.91
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38870-89-2
108.52
JJKWHOSQTYYFAE-UHFFFAOYSA-N
96623
COCC(=O)Cl
C3H5ClO2
MFCD00000728
methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
2-methoxyacetyl chloride

Specifications

38870-89-2
1.1870g/mL
28°C
97%
C3H5ClO2
CH3OCH2COCl
50 g
1.187
Solubility in water: reacts
COCC(=O)Cl
108.52
96623
97%
Methoxyacetyl chloride, Stabilized
Colorless to Yellow
112°C to 113°C
Authentic
Glass bottle
1.4190 to 1.422
MFCD00000728
12,310
methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
JJKWHOSQTYYFAE-UHFFFAOYSA-N
2-methoxyacetyl chloride
0.3%mgO
108.52
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 254-169-2

SDS
Documents

Documents

RUO – Research Use Only