missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyacetic Acid 96.0+%, TCI America™
$35.41 - $197.59
Chemical Identifiers
| CAS | 625-45-6 |
|---|---|
| Molecular Formula | C3H6O3 |
| Molecular Weight (g/mol) | 90.078 |
| MDL Number | MFCD00004308 |
| InChI Key | RMIODHQZRUFFFF-UHFFFAOYSA-N |
| Synonym | methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 |
| PubChem CID | 12251 |
| IUPAC Name | 2-methoxyacetic acid |
| SMILES | COCC(=O)O |
Chemical Identifiers
| 625-45-6 | |
| 90.078 | |
| RMIODHQZRUFFFF-UHFFFAOYSA-N | |
| 12251 | |
| COCC(=O)O |
| C3H6O3 | |
| MFCD00004308 | |
| methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 | |
| 2-methoxyacetic acid |
Specifications
| 625-45-6 | |
| 204°C | |
| MFCD00004308 | |
| 3265 | |
| RMIODHQZRUFFFF-UHFFFAOYSA-N | |
| 2-methoxyacetic acid | |
| 12251 | |
| ≥96.0% (T) | |
| Methoxyacetic Acid |
| Yellow | |
| C3H6O3 | |
| 25 g | |
| methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 | |
| COCC(=O)O | |
| 90.078 | |
| 90.08 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3484
RTECSNumber : AI8650000
TSCA : Yes