missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetic Acid 96.0+%, TCI America™

Catalog No. M0827500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0827500G 500 g
M082725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0827500G Supplier TCI America Supplier No. M0827500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-45-6
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00004308
InChI Key RMIODHQZRUFFFF-UHFFFAOYSA-N
Synonym methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
PubChem CID 12251
IUPAC Name 2-methoxyacetic acid
SMILES COCC(=O)O

Specifications

Color Yellow
Boiling Point 204°C
Quantity 500 g
UN Number 3265
Formula Weight 90.08
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material Methoxyacetic Acid
EINECSNumber (2)-3484
RTECSNumber AI8650000
TSCA Yes