Learn More
Methoxyacetic acid, 97%
CAS: 625-45-6 | C3H6O3 | 90.078 g/mol
$66.56 - $460.74
Chemical Identifiers
| CAS | 625-45-6 |
|---|---|
| Molecular Formula | C3H6O3 |
| Molecular Weight (g/mol) | 90.078 |
| MDL Number | MFCD00004308 |
| InChI Key | RMIODHQZRUFFFF-UHFFFAOYSA-N |
| Synonym | methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 |
| PubChem CID | 12251 |
| IUPAC Name | 2-methoxyacetic acid |
| SMILES | COCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2291718
|
Thermo Scientific Chemicals
B2291718 |
50 g |
Each for $66.56
|
|
|||||
|
AAB2291730
|
Thermo Scientific Chemicals
B2291730 |
250 g |
Each for $167.70
|
|
|||||
|
AAB229170B
|
Thermo Scientific Chemicals
B229170B |
1000 g |
Each for $460.74
|
|
|||||
Description
Methoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethoxyacetic acid is used in zebrafish embryotoxicity test to evaluate chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis. It is also used in industry as gelling, viscosity and stabilizer reagent.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
| 625-45-6 | |
| 90.078 | |
| RMIODHQZRUFFFF-UHFFFAOYSA-N | |
| 12251 | |
| COCC(=O)O |
| C3H6O3 | |
| MFCD00004308 | |
| methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 | |
| 2-methoxyacetic acid |
Specifications
| 625-45-6 | |
| 1.174 | |
| 202°C to 204°C | |
| Odorless | |
| 1.4165 | |
| MFCD00004308 | |
| UN3265 | |
| methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 | |
| RMIODHQZRUFFFF-UHFFFAOYSA-N | |
| 2-methoxyacetic acid | |
| 12251 | |
| 97% |
| 8°C | |
| 1.6 | |
| 114°C (237°F) | |
| C3H6O3 | |
| CH3OCH2CO2H | |
| 50 g | |
| 635800 | |
| Miscible with water. | |
| COCC(=O)O | |
| 90.078 | |
| 90.08 | |
| Methoxyacetic acid |
Safety and Handling
GHS H Statement
H360-H314-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335-H360FD
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 210-894-6
RTECSNumber : AI8650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only