missing translation for 'onlineSavingsMsg'
Learn More

Methoxy(dimethyl)octadecylsilane 90.0+%, TCI America™

Catalog No. p-7133535
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30115ML 5 mL
M301125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30115ML Supplier TCI America Supplier No. M30115ML
Only null left

Chemical Identifiers

CAS 71808-65-6
Molecular Formula C21H46OSi
Molecular Weight (g/mol) 342.68
MDL Number MFCD00042920
InChI Key RUFRLNPHRPYBLF-UHFFFAOYSA-N
PubChem CID 172903
IUPAC Name methoxydimethyloctadecylsilane
SMILES CCCCCCCCCCCCCCCCCC[Si](C)(C)OC

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 342.68
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxy(dimethyl)octadecylsilane