missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methocarbamol 98.0+%, TCI America™
$55.92 - $167.76
Chemical Identifiers
| CAS | 532-03-6 |
|---|---|
| Molecular Formula | C11H15NO5 |
| Molecular Weight (g/mol) | 241.243 |
| MDL Number | MFCD00057662 |
| InChI Key | GNXFOGHNGIVQEH-UHFFFAOYSA-N |
| Synonym | Carbamic Acid 2-Hydroxy-3-(2-methoxyphenoxy)propyl Ester, 2-Hydroxy-3-(2-methoxyphenoxy)propyl Carbamate |
| PubChem CID | 4107 |
| ChEBI | CHEBI:77498 |
| IUPAC Name | [2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate |
| SMILES | COC1=CC=CC=C1OCC(COC(=O)N)O |
Chemical Identifiers
| 532-03-6 | |
| 241.243 | |
| GNXFOGHNGIVQEH-UHFFFAOYSA-N | |
| 4107 | |
| [2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate |
| C11H15NO5 | |
| MFCD00057662 | |
| Carbamic Acid 2-Hydroxy-3-(2-methoxyphenoxy)propyl Ester, 2-Hydroxy-3-(2-methoxyphenoxy)propyl Carbamate | |
| CHEBI:77498 | |
| COC1=CC=CC=C1OCC(COC(=O)N)O |
Specifications
| 96°C | |
| White | |
| MFCD00057662 | |
| GNXFOGHNGIVQEH-UHFFFAOYSA-N | |
| [2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate | |
| 241.243 | |
| CHEBI:77498 | |
| ≥98.0% (HPLC,N) | |
| Methocarbamol |
| 532-03-6 | |
| C11H15NO5 | |
| Carbamic Acid 2-Hydroxy-3-(2-methoxyphenoxy)propyl Ester, 2-Hydroxy-3-(2-methoxyphenoxy)propyl Carbamate | |
| COC1=CC=CC=C1OCC(COC(=O)N)O | |
| 5 g | |
| 4107 | |
| 241.24 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0293&(9)-1471
RTECSNumber : TY8750000
TSCA : Yes