missing translation for 'onlineSavingsMsg'
Learn More

Methionine, USP, 98.5-101.5%, Spectrum™ Chemical

M1441, 63-68-3, C5H11NO2S

Supplier:  Spectrum Chemical Mfg Cor M14411KG

 View more versions of this product

Catalog No. 18604387

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Methionine, USP is used to prevent liver damage in acetaminophen poisoning. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

63-68-3
0.4%
Amber Glass Bottle
MFCD00063097 MFCD00801344
+22.4° to +24.7°
CSCC[C@H](N)C(O)=O
149.21
USP
100%
0.3%
C5H11NO2S
1 kg
FFEARJCKVFRZRR-BYPYZUCNSA-N
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
98.5 to 101.5%

Chemical Identifiers

63-68-3
149.21
FFEARJCKVFRZRR-BYPYZUCNSA-N
CSCC[C@H](N)C(O)=O
C5H11NO2S
MFCD00063097 MFCD00801344
(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions