missing translation for 'onlineSavingsMsg'
Learn More

Methionine, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18604384
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604384 25 g Amber Glass Bottle
18604385 100 g Amber Glass Bottle
18604386 500 g Amber Glass Bottle
18604387 1 kg Amber Glass Bottle
18604388 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604384 Supplier Spectrum Chemical Mfg Cor Supplier No. M144125GM
Only null left

M1441, 63-68-3, C5H11NO2S

Spectrum™ Chemical Methionine, USP is used to prevent liver damage in acetaminophen poisoning. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
SMILES CSCC[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.4%
Loss on Drying 0.3%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +22.4° to +24.7°
Percent Purity 98.5 to 101.5%
Grade USP