missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betamethasone, 97%

Catalog No. AC459070250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459070250 25 g
AC459070010 1 g
AC459070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC459070250 Supplier Thermo Scientific Chemicals Supplier No. 459070250

missing translation for 'validMainframeMsg'

CAS: 378-44-9 | C22H29FO5 | 392.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 378-44-9
Molecular Formula C22H29FO5
Molecular Weight (g/mol) 392.46
InChI Key UREBDLICKHMUKA-DVTGEIKXSA-N
Synonym betamethasone, betadexamethasone, flubenisolone, celestone, betamethazone, rinderon, betacorlan, betacortril, betamamallet, betametasone
PubChem CID 9782
ChEBI CHEBI:3077
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)CO)O)C)O)F)C

Specifications

Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 392.46
Percent Purity 97%
Chemical Name or Material βmethasone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
EINECSNumber 206-825-4

RUO – Research Use Only