missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Synthetic (Methyl Alcohol), Reagents

Catalog No. 01337524
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337524 1 L
01337523 200 L
01337522 20 L
01337521 4 L
01337520 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337524 Supplier Labchem Inc Supplier No. C21210001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methanol, Synthetic (Methyl Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7866
Boiling Point 64.5°C
Grade Reagent
Chemical Name or Material Methanol, Synthetic (Methyl Alcohol)