missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Supelco

Catalog No. M1047322000
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1047322000 2 L
M1047320500 500 mL
01257784 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1047322000 Supplier Merck Emd Millipore Supplier No. 1047322000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tested for PFAS Methods LiChrosolv™

  • Application Security: Quality control on 40 PFAS analytes according to EPA 533, EPA 537.1, and EPA 1633.
  • Time and Cost Savings: EPA 1633 requires batch validation unless the vendor tests for PFAS analytes listed in this method.
  • Lowest Impurity Profile: Ensures an interference-free baseline.
  • LC-MS Suitability: ESI/APCI (+) < 2 ppb; ESI/APCI (-) < 10 ppb.
  • Minimized Contamination: Borosilicate glass bottles reduce contamination with metal ions.
  • Trace Metal Impurity Control: Levels of trace metal impurities are minimized to <5 ppb, reducing metal ion adduct formation.
  • Microfiltration: Solvents are filtered through a 0.2 μm filter to ensure purity.

Chemical Identifiers

Molecular Weight (g/mol) 32.04

Specifications

CAS 67-56-1
Quantity 2 L
Molecular Formula CH4O
Boiling Point 64.7°C (lit.)
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Percent Purity ≥99.9% (GC)
Chemical Name or Material Methanol