missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8% (GC)
Supplier: Honeywell-Riedel-de Haen 322134X25L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Methanol | |
| Reag. Ph. Eur. reag. ISO | |
| Colorless | |
| 0.791g/mL at 25°C | |
| 4 x 2.5 L | |
| 9°C (48.2°F) | |
| Poly Bottle | |
| 0 to 0.0005 % | |
| CH4O | |
| CH3OH | |
| 410mmHg (50°C), 97.68mmHg (20°C) | |
| 1098229 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04g/mol | |
| ACS/puriss. p.a. |
| -98°C | |
| 67-56-1 | |
| Liquid | |
| 64°C to 65°C | |
| 0.36 | |
| 725°F | |
| 1.328 to 1.33 | |
| Free acid (as HCOOH): ≤0.002% | |
| Free Alkali (as NH3): ≤0.00005% | |
| MFCD00004595 | |
| UN1230 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.8% (GC) |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Safety and Handling
P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
H225-H301 + H311 + H331-H370
DOTInformation : Class 3 - PG 2 - Methanol
EINECSNumber : 200-659-6
RTECSNumber : PC1400000