missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Optima™ LC/MS Grade, Fisher Chemical

Catalog No. A456500
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A456500 500 mL
A4561 1 L
A456212 2.5 L
A4564 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A456500 Supplier Thermo Fisher Scientific Supplier No. A456 500
Only null left

Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol | CAS: 67-56-1 | CH4O | 32.04 g/mol

Methanol, >-99.9%, CH4O, CAS Number-67-56-1, wood alcohol, methyl hydroxide, colonial spirit, columbian spirit, wood naphtha, carbinol, methyl alcohol, pyroxylic spirit, wood spirit, methylol

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.2 AU at 220nm, ≤0.4 AU at 214nm, ≤0.5 AU at 210nm
Color Colorless
Density 0.791g/cm³
Boiling Point 64.7°C
Quantity 500 mL
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤1ppm
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid ≤0.0003meq/g
Titratable Base ≤0.0002meq/g
Linear Formula CH3OH
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55cp at 20°C
Water ≤0.02%
Merck Index 15, 6029
Formula Weight 32.04
Percent Purity ≥99.9%
Grade LC-MS & UHPLC-UV
Assay GC
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health: 1
Flammability: 3
Instability: 0
DOTInformation DOT Class 3, : Flammable Liquid