missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, OmniSolv™, Pesticide Residue Grade, MilliporeSigma™
$301.04 - $301.04
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
| PubChem CID | 887 |
| ChEBI | CHEBI:17790 |
| IUPAC Name | methanol |
| SMILES | CO |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Specifications
| 67-56-1 | |
| 0.792g/cm3 (20°C) | |
| 6 to 36.5% | |
| Glass Bottle | |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| Methanol, For Pesticide Residue Grade; also for Gas Chromatography |
| Colorless | |
| 4 L | |
| 464°C / 867.2°F | |
| Liquid | |
| CH3OH | |
| 12.9 kPa (20°C) | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.05 |
Safety and Handling
Recommended Storage : Room Temperature