missing translation for 'onlineSavingsMsg'
Learn More

Methanol, OmniSolv™, Pesticide Residue Grade, MilliporeSigma™

Catalog No. MMX04841
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX04841 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX04841 Supplier Technidata Supplier No. US1MX0484/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Density 0.792g/cm3 (20°C)
Quantity 4 L
Explosion Limit 6 to 36.5%
Ignition Point 464°C / 867.2°F
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 12.9 kPa (20°C)
Linear Formula CH3OH
Formula Weight 32.05
Chemical Name or Material Methanol, For Pesticide Residue Grade; also for Gas Chromatography
Show More Show Less
Recommended Storage Room Temperature