missing translation for 'onlineSavingsMsg'
Learn More

Methanol, OmniSolv Gradient Grade, MilliporeSigma™

Catalog No. MMX04886
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Glass Bottle
NOWPak™
Poly-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX04886 1 L Glass Bottle
MMX04881 4 L Glass Bottle
MMX0488P1 4 L Poly-coated Glass Bottle
MMX0488NP20 20 L NOWPak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MMX04886 Supplier Technidata Supplier No. US1MX0488/6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -97.8°C
Color Colorless
Density 0.792g/cm3 (20°C)
Quantity 1 L
Explosion Limit 5.5 to 44%(V)
Flash Point Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F)
Ignition Point 464°C (867.2°F)
Odor Alcohol-like
Packaging Glass Bottle
Linear Formula CH3OH
Vapor Pressure 12.9kPa (97mm Hg) [20°C]
UN Number UN 1230
Boiling Point 64.5°C
Formula Weight 32.04
Percent Purity ≥99.9%
Chemical Name or Material Methanol
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Room Temperature