missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol LM Prime, Transene
Methanol Semiconductor Grade
$722.40 - $5888.13
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
50-274-4715
|
Doe and Ingalls Management LLC
T060METHANOLM55GAL |
55 Gallon |
Each for $5,888.13
|
|
|||||
|
50-274-4716
|
Doe and Ingalls Management LLC
T0701355GAL |
1 gal. |
Case of 4 Each for $722.40
|
|
|||||
Description
Specifically formulated for electronics manufacturing are referred to as electronic grade solvents. These solvents contain very low levels of heavy metals, typically measured in parts per billion or parts per trillion. Achieving the required purity levels involves several stages of purification. They are commonly utilized as cleaners and etchants in the production of semiconductors and microprocessors.
- Common Uses and Applications:
- HPLC (High-Performance Liquid Chromatography)
- Reagent
- Electronics
- Cleaning agent
- Semiconductor synthesis
- Solvent
- Etching agent
Safety and Shipping Information:DOT: Methanol, 3, UN1230, PG II
Chemical Identifiers
| 67-56-1 | |
| 32 g/mol |
Specifications
| 67-56-1 | |
| 55 Gallon | |
| 32 g/mol | |
| Electronic |
| 64.7°C | |
| HDPE | |
| 99.90% | |
| Methyl Alcohol |