missing translation for 'onlineSavingsMsg'
Learn More

Methanol LM Prime, Transene

Catalog No. 502744715
Change view
Click to view available options
Quantity:
55 Gallon
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502744715 55 Gallon
502744716 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 502744715 Supplier Doe and Ingalls Management LLC Supplier No. T060METHANOLM55GAL

Methanol Semiconductor Grade

Specifically formulated for electronics manufacturing are referred to as electronic grade solvents. These solvents contain very low levels of heavy metals, typically measured in parts per billion or parts per trillion. Achieving the required purity levels involves several stages of purification. They are commonly utilized as cleaners and etchants in the production of semiconductors and microprocessors.

  • Common Uses and Applications:
  • HPLC (High-Performance Liquid Chromatography)
  • Reagent
  • Electronics
  • Cleaning agent
  • Semiconductor synthesis
  • Solvent
  • Etching agent

Safety and Shipping Information:DOT: Methanol, 3, UN1230, PG II

Chemical Identifiers

CAS 67-56-1
Molecular Weight (g/mol) 32 g/mol

Specifications

Quantity 55 Gallon
Packaging HDPE
Boiling Point 64.7°C
Percent Purity 99.90%
Grade Electronic
Chemical Name or Material Methyl Alcohol