missing translation for 'onlineSavingsMsg'
Learn More

Methanol, LC-MS CHROMASOLV™, Solstice

Catalog No. 6000954
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000954 4 x 2.5 L
6000952 1 L
6000953 2.5 L
6000956 6 x 1 L
6000955 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6000954 Supplier Solstice Supplier No. 349664X2.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Linear Formula CH3OH
Vapor Pressure 410 mmHg (50°C), 97.68 mmHg (20°C)
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade LC-MS CHROMASOLV
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
RTECSNumber PC1400000