missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Laboratory Reagent, ≥99.6%, Honeywell™
Laboratory Reagent,≥99.6%
Supplier: Honeywell Chemicals 1799574X25L
Description
- ≥95% for general laboratory use.
Specifications
| Methanol | |
| 67-56-1 | |
| 0.791g/mL at 25°C | |
| 4 x 2.5 L | |
| 9°C (48.2°F) | |
| Glass Bottle | |
| n20/D 1.329(lit.) | |
| CH3OH | |
| 97.68mmHg (20°C), 410mmHg (50°C) | |
| 1098229 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04g/mol | |
| Laboratory Reagent |
| -98°C | |
| Colorless | |
| 65°C | |
| 0.36 | |
| 725°F | |
| Liquid | |
| CH4O | |
| MFCD00004595 | |
| UN1230 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.6% |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Safety and Handling
P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
H225-H301 + H311 + H331-H370
DOTInformation : class 3 - PG 2 - Methanol
EINECSNumber : 200-659-6
RTECSNumber : PC1400000