missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Laboratory Reagent, ≥99.6%, Solstice

Catalog No. 60045005
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
4 x 4 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045005 2.5 L Glass Bottle
60045008 4 x 2.5 L Glass Bottle
60045009 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60045005 Supplier Solstice Supplier No. 1799572.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory Reagent,≥99.6%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.329(lit.)
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number UN1230
Beilstein 1098229
Boiling Point 65°C
Formula Weight 32.04g/mol
Percent Purity ≥99.6%
Grade Laboratory Reagent
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
Precautionary Statement H225-H301 + H311 + H331-H370
DOTInformation class 3 - PG 2 - Methanol
EINECSNumber 200-659-6
RTECSNumber PC1400000