missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, HPLC Plus, ≥99.9%; Honeywell™ Riedel-de Haën™
CHROMASOLV™ Plus, for HPLC
$1195.70 - $1195.70
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| PubChem CID | 887 |
| ChEBI | CHEBI:17790 |
| IUPAC Name | methanol |
| SMILES | CO |
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| CHEBI:17790 | |
| CO |
| CH4O | |
| MFCD00004595 | |
| 887 | |
| methanol |
Specifications
| 67-56-1 | |
| 4 x 4 L | |
| 725°C | |
| CH4O | |
| MFCD00004595 | |
| UN1230 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.9% | |
| Methanol |
| 0.791g/mL at 25°C, 0.791g/mL at 25°C | |
| 0.36 | |
| Glass Bottle | |
| CH3OH | |
| 410mmHg (50°C), 97.68mmHg (20°C) | |
| 1098229 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04g/mol | |
| HPLC Plus |
Safety and Handling
P210-P260-P280-P301 + P310-P311
DOTInformation : class 3 - PG 2 - Methanol
RTECSNumber : 200-659-6