missing translation for 'onlineSavingsMsg'
Learn More

Methanol (Histological), Fisher Chemical™

Catalog No. p-22324
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A433P4 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A433P4 Supplier Thermo Fisher Scientific Supplier No. BPA433P4
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in histology applications

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791g/cm³
Quantity 4 L
Identification Pass Test
Packaging Poly Bottle
Physical Form Liquid
Linear Formula CH3OH
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
UN Number 1230
Boiling Point 64.7°C
Formula Weight 32.04
Grade Histological
Chemical Name or Material Methanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
DOTInformation DOT Class 3, : Flammable Liquid