missing translation for 'onlineSavingsMsg'
Learn More

Methanol, GC2™ , for GC, pesticide residue analysis, >99.9%, Honeywell Burdick & Jackson

Catalog No. 6000933
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000933 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000933 Supplier Honeywell Chemicals Supplier No. GC2304
Only null left

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Quantity 4 x 4 L
Flash Point 11°C
Packaging Glass Bottle
Water 0.1% max.
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Solubility Information Miscible in Water at 20°C
Formula Weight 32.04
Percent Purity >99.9%
Grade GC/Pesticide Residue Analysis
Chemical Name or Material Methanol, GC2™ , for GC, pesticide residue analysis, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol