missing translation for 'onlineSavingsMsg'
Learn More

Methanol, For HPLC analysis, MilliporeSigma™

Catalog No. MMX0475P1
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX0475P1 4 L Poly-coated Glass Bottle
MMX04751 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MMX0475P1 Supplier Technidata Supplier No. MX0475P1
Only null left

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Quantity 4 L
Packaging Poly-coated Glass Bottle
Physical Form Liquid
Linear Formula CH3OH
Formula Weight 32.4
Grade ACS/HPLC/NF/USP
Chemical Name or Material Methanol, For HPLC
Recommended Storage Room Temperature