missing translation for 'onlineSavingsMsg'
Learn More

Methanol For gas chromatography MS SupraSolv™, MilliporeSigma™

Catalog No. M1008372500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1008372500 2.5 L Glass Bottle
M1008371000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1008372500 Supplier Technidata Supplier No. 1.00837.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For gas chromatography MS SupraSolv™

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 2.5 L
Flash Point 10°C (50°F)
Ignition Point 455°C (851°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 128 hPa (20°C)
UN Number 1230
Boiling Point 64.5°C
Solubility Information Soluble in water at 20°C
Formula Weight 32.04
Chemical Name or Material Methanol
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
Precautionary Statement H370: Causes damage to organs
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.