missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™
Specifications
Specifications
| Melting Point | -98°C |
| Color | Colorless |
| Density | 0.79g/cm3 at 20°C |
| Quantity | 1 L |
| Flash Point | 10°C (50°F) |
| Ignition Point | 455°C (851°F) |
| Odor | Charactristic |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 128 hPa (20°C) |
| Show More |
Safety and Handling
| Hazard Category | H225: Highly flammable liquid and vapour. |
| Hazard Statement | H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled |
| Precautionary Statement | H370: Causes damage to organs |
| Recommended Storage | 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons. |