Learn More
Methanol, Environmental Grade, 99.8+%
CAS: 67-56-1 | CH4O | 32.04 g/mol
Supplier: Thermo Scientific Chemicals 040980K7
Description
It is suitable for environmental and GC analysis. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. Used for producing bio-diesel, synthesis of formaldehyde and acetic acid, useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin. It is useful for the preparation of compact direct methanol fuel cells for portable application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methanol | |
| Environmental Grade | |
| 0.791 g/mL | |
| 4 L | |
| Liquid | |
| >99.8% | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04 |
| °C to 98°C | |
| 67-56-1 | |
| 64.7 | |
| 11°C (52°F) | |
| 1.329 | |
| CH4O | |
| 1098229 | |
| Miscible in water. | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| Environmental Plus |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370-H372
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 200-659-6
RTECSNumber : PC1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only