missing translation for 'onlineSavingsMsg'
Learn More

Methanol (Electronic), Fisher Chemical

Catalog No. A9474
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A9474 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A9474 Supplier Thermo Fisher Scientific Supplier No. BPA9474
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791g/cm³
Quantity 4 L
Identification Pass Test
Packaging Poly Bottle
Physical Form Liquid
Residue after Evaporation 10ppm max.
Assay Percent Range 99.8% min by GC
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0002mEq/g max.
Vapor Pressure 128 hPa at 20°C
Linear Formula CH3OH
Viscosity 0.55 cP at 20°C
Water 0.1% max.
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity ≥99.8%
Grade For use in electronics and circuit board manufacturing
Chemical Name or Material Methanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0