missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Product filtered through a 0.2μm filter
Supplier: Honeywell-Chromasolv 3486025L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
| Methanol | |
| for HPLC | |
| 0.791g/mL at 25°C | |
| 2.5 L | |
| CH4O | |
| MFCD00004595 | |
| 1098229 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04g/mol | |
| CHROMASOLV |
| -98°C | |
| 67-56-1 | |
| 64.7°C | |
| n20/D 1.329 | |
| CH3OH | |
| 97.68mmHg (20°C) | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.9% |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Safety and Handling
P210-P260-P280-P301 + P310-P311
H225-H301 + H311 + H331-H370
EINECSNumber : 200-659-6
RTECSNumber : PC1400000