missing translation for 'onlineSavingsMsg'
Learn More

Methanol, For pesticide residue analysis, Honeywell Riedel-de Haën™

Catalog No. p-7072100
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000957 1 L
6000958 2.5 L
60046346 4 x 2.5 L
6000963 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000957 Supplier Honeywell Chemicals Supplier No. 344851L
Only null left

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Linear Formula CH3OH
Vapor Pressure 410 mmHg (50°C), 97.68 mmHg (20°C)
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.9% (GC)
Grade CHROMASOLV
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
RTECSNumber PC1400000