missing translation for 'onlineSavingsMsg'
Learn More

Solstice Advanced METHANOL BIOSYN 2LT

Supplier:  Honeywell Chemicals 4944372L

METHANOL BIOSYN 2LT

Catalog No. 60046979


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methanol
67-56-1
CH3OH
UN1230
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
≥99.93%
65°C
0.36
725°F
Glass Bottle
410mmHg (50°C), 97.68mmHg (20°C)
-98°C
CH4O
MFCD00004595
1098229
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04g/mol
0.791g/mL at 25°C
2 L
9°C (48.2°F)
Characteristic
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions