missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Biograde, 99.8+%

CAS: 67-56-1 | CH4O | 32.04 g/mol

$160.81 - $496.05

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44571K7
View Documents
Thermo Scientific Chemicals
044571K7
4 L
Each for $160.81
Only null left
 
AA44571M6
View Documents
Thermo Scientific Chemicals
044571M6
4 x 4 L
Each for $496.05
Only null left
 
Description

Description

It is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

-98°C
0.791 g/mL
4 L
Liquid
>99.8%
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Methanol, Filtered through 0.2μfilter
67-56-1
64.7°C
11°C (52°F)
1.329
CH4O
1098229
Miscible in water.
CO
32.04
CHEBI:17790
Biochemical
Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331-H370-H372

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 200-659-6

RTECSNumber : PC1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only