missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Purge and Trap grade, for VOC Analysis using Purge and Trap methods, >99.9%, Solstice
Description
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Specifications
Specifications
| CAS Max % | 100.00 |
| Density | 0.792 g/cm3 |
| Quantity | 4 x 4 L |
| Flash Point | 11°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Boiling Point | 64.7°C |
| Grade | HPLC |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | H370+H225 |
| Precautionary Statement | P210+P233+P240+P241+P242+P243+P260+P264+P270+P280 |
| Recommended Storage | Room Temperature |