missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9%, Honeywell Burdick & Jackson
Supplier: Honeywell Inc LC23025
Specifications
| Methanol | |
| 67-56-1 | |
| 0.792 g/cm3 | |
| 4 x 2.5 L | |
| GL45 Glass Bottle | |
| MFCD00004595 | |
| 0.05% max. | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04 | |
| LC-MS |
| B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9% | |
| 100.0000% | |
| 64.7°C | |
| 11°C | |
| CH4O | |
| UN1230 | |
| 1098229 | |
| Miscible in Water at 20°C | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| >99.9% |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol