missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, B&J Brand™, for HPLC, for spectrophotometry, >99.9%, Honeywell Burdick & Jackson
$3693.58 - $5597.65
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
| PubChem CID | 887 |
| ChEBI | CHEBI:17790 |
| IUPAC Name | methanol |
| SMILES | CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6000971
|
Honeywell-Burdick and Jackson
CS23056 |
56 L | Returnable Steel Drum |
Each for $3,693.58
|
|
||||
|
6000966
|
Honeywell-Burdick and Jackson
CS230200 |
200 L | Returnable Steel Drum |
Each for $5,597.65
|
|
||||
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Specifications
| 67-56-1 | |
| 100.0000% | |
| 64.7°C | |
| 11°C | |
| CH4O | |
| UN1230 | |
| 1098229 | |
| Miscible in Water at 20°C | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| >99.9% | |
| Methanol, B&J Brand™, for HPLC, for spectrophotometry, >99.9% |
| 0.16 AU max. at 225 nm, 0.02 AU max. at 250 nm, 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm | |
| 0.792 g/cm3 | |
| 56 L | |
| Returnable Steel Drum | |
| MFCD00004595 | |
| 0.05% max. | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04 | |
| HPLC |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol