missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Anhydrous, DriSolv™, Septum-Sealed, MilliporeSigma™

Catalog No. MMX04726
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX04726 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX04726 Supplier Technidata Supplier No. US1MX0472/6

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Packed under Nitrogen

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Quantity 1 L
Physical Form Liquid Free From Particulates
Residue after Evaporation 1ppm max.
Assay Percent Range 99.8% min.
Linear Formula CH3OH
Water 50ppm max.
Formula Weight 32.04
Chemical Name or Material Methanol, Septum-sealed
Recommended Storage Room Temperature