missing translation for 'onlineSavingsMsg'
Learn More

Methanol, AMD CHROMASOLV™, ≥99.9%, Solstice

Catalog No. 6000934
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000934 2.5 L Glass Bottle
60046504 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000934 Supplier Honeywell Inc Supplier No. 348982.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ AMD,≥99.9%

  • Solvents designed for high-performance thin-layer chromatography

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C (literature)
CAS Max % ≤100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Quantity 2.5 L
Flash Point 11°C
Packaging Glass Bottle
Linear Formula CH3OH
UN Number UN1230
Beilstein 1098229
Boiling Point 64°C to 65°C
Formula Weight 32.04
Percent Purity >99.9%
Grade High Purity
Chemical Name or Material Methanol, AMD CHROMASOLV™, ≥99.9%
Show More Show Less
ShelfLife 1800 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
EINECSNumber 200-659-6
Recommended Storage Room Temp