missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%, Solstice

Catalog No. 6000979
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000979 204 L Metal Drum
6000980 4 x 4 L Glass Bottle
6000981 4 x 4 L Poly Bottle
6000978 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6000979 Supplier Solstice Supplier No. LP230204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Quantity 204 L
Flash Point 11°C
Packaging Metal Drum
Water 0.1% max.
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Solubility Information Miscible in Water at 20°C
Formula Weight 32.04
Percent Purity >99.8%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Methanol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol