missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.9%, Honeywell™

Catalog No. p-7112007
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000976 4 x 4 L Glass Bottle
6000977 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000976 Supplier Honeywell Inc Supplier No. AH2304
Only null left

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Boiling Point 64.7°C
Quantity 4 x 4 L
Flash Point 11°C
Packaging Glass Bottle
UN Number UN1230
Beilstein 1098229
Water 0.05% max.
Solubility Information Miscible in Water at 20°C
Formula Weight 32.04
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Methanol, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
EINECSNumber 200-659-6