missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), Reagents
Specifications
Specifications
| Melting Point | -98°C |
| Color | Colorless |
| Quantity | 20 L |
| Packaging | Steel Pail |
| Physical Form | Liquid |
| Specific Gravity | 0.7866 |
| Boiling Point | 64.5°C |
| Grade | ACS |
| Chemical Name or Material | Methanol, ACS Reagent, Absolute (Methyl Alcohol, Anhydrous) |