Learn More
Methanol, ACS Grade, ≥ 99.8%, LabChem™
$57.43 - $207.00
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
| PubChem CID | 887 |
| ChEBI | CHEBI:17790 |
| IUPAC Name | methanol |
| SMILES | CO |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Specifications
| -98°C | |
| Colorless | |
| ≤0.0003meq/gcSt | |
| Carbon dioxide; Carbon monoxide | |
| 65°C | |
| 9.7°C | |
| Amber Glass | |
| ≤0.001% | |
| CH4O | |
| MFCD00004595 | |
| ≤0.1% | |
| Soluble in water | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.8% | |
| Methanol |
| 67-56-1 | |
| 100 | |
| ≤0.0002meq/gcSt | |
| 792kg/m3 | |
| 500 mL | |
| Passes Test | |
| Liquid | |
| 792kg/m3 | |
| CH3OH | |
| UN1230 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04 | |
| ACS |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (liver, kidneys, central nervous system, optic nerve) (dermal, oral).
GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
EINECSNumber : 200-659-6
Recommended Storage : Room Temperature