missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Methanol | |
| 67-56-1 | |
| 0.79g/cm3 at 20°C | |
| Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F) | |
| Alcohol-like | |
| Liquid | |
| CH3OH | |
| UN 1230 | |
| Soluble in water | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.9% |
| -97.8°C | |
| Colorless | |
| 20 L | |
| 464°C (867.2°F) | |
| Metal Can | |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04 | |
| ACS/NF/USP |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Room Temperature