missing translation for 'onlineSavingsMsg'
Learn More

Methanol ACS, MilliporeSigma™

Catalog No. MMX0485P4
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
Poly-coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX0485P4 4 L Poly-coated Glass Bottle
MMX04856 500 mL Glass Bottle
MMX04857 4 L Glass Bottle
361020510 4 L Poly Bottle
MMX04855 20 L Metal Can
MMX048520 200 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MMX0485P4 Supplier Technidata Supplier No. MX0485P4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -97.8°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 4 L
Flash Point Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F)
Ignition Point 464°C (867.2°F)
Odor Alcohol-like
Packaging Poly-coated Glass Bottle
Physical Form Liquid
Linear Formula CH3OH
UN Number UN 1230
Solubility Information Soluble in water
Formula Weight 32.04
Percent Purity ≥99.9%
Grade ACS/NF/USP
Chemical Name or Material Methanol
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Room Temperature